siRNA Details
| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106428.2 | NP_001099898.2 | 8 | 1448 | ||
| XM_006232297.2 | XP_006232359.1 | 8 | 1535 | |||
| XM_006232298.2 | XP_006232360.1 | 8 | 949 | |||
| GenBank mRNA | BC168157.1 | - | 8 | 1448 | 8363916 | |

Hspa4l
Genomic Map