siRNA Details
| Gene Symbol: Slc20a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017223.2 | NP_058919.1 | 2 | 500 | ||
| XM_008771351.1 | XP_008769573.1 | 2 | 1507 | |||
| XM_008771352.1 | XP_008769574.1 | 2 | 1147 | |||
| GenBank mRNA | BC070908.1 | - | 2 | 500 | 7103640 | |
| L19931.1 | - | 1 | 356 | |||

Slc20a2
Genomic Map