siRNA Details
| Gene Symbol: Slc22a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019230.1 | NP_062103.1 | 6 | 1421 | ||
| GenBank mRNA | AF055286.1 | - | 6 | 1421 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a3| Gene Symbol: Slc22a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019230.1 | NP_062103.1 | 6 | 1421 | ||
| GenBank mRNA | AF055286.1 | - | 6 | 1421 | ||