siRNA Details
| Gene Symbol: Schip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100666.1 | NP_001094136.1 | 5, 6 | 1058 | ||
| XM_008761079.1 | XP_008759301.1 | 2, 3 | 3996 | |||
| XM_008761080.1 | XP_008759302.1 | 2, 3 | 865 | |||
| XM_008761081.1 | XP_008759303.1 | 2, 3 | 741 | |||
| XM_008761082.1 | XP_008759304.1 | 1, 2 | 3013 | |||
| XM_008761083.1 | XP_008759305.1 | 1, 2 | 312 | |||
| GenBank mRNA | BC092618.1 | - | 1, 2 | 30 | ||
| BC169016.1 | - | 1, 2 | 259 | |||

Schip1
Genomic Map