siRNA Details
| Gene Symbol: Etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198742.2 | NP_942037.2 | 2, 3 | 351 | ||
| XM_006232507.1 | XP_006232569.1 | 2, 3 | 351 | |||
| XM_008761084.1 | XP_008759306.1 | 3, 4 | 577 | |||
| GenBank mRNA | AY365022.1 | - | 2, 3 | 177 | ||
| BC081890.1 | - | 2, 3 | 351 | 7109511 | ||
| FQ211230.1 | - | 2, 3 | 409 | |||
| FQ218772.1 | - | 3, 4 | 582 | |||

Etfdh
Genomic Map