siRNA Details
| Gene Symbol: Etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198742.2 | NP_942037.2 | 5 | 751 | ||
| XM_006232507.1 | XP_006232569.1 | 5 | 751 | |||
| XM_008761084.1 | XP_008759306.1 | 6 | 977 | |||
| GenBank mRNA | AY365022.1 | - | 5 | 577 | ||
| BC081890.1 | - | 5 | 751 | 7109511 | ||
| FQ211230.1 | - | 5 | 808 | |||
| FQ218772.1 | - | 6 | 982 | |||

Etfdh
Genomic Map