siRNA Details
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201417.1 | NP_958820.1 | 16 | 1724 | ||
XM_006232508.2 | XP_006232570.1 | 16 | 1861 | |||
XM_006232510.2 | XP_006232572.1 | 13 | 1415 | |||
XM_008761085.1 | XP_008759307.1 | 12 | 3165 | |||
XM_008761086.1 | XP_008759308.1 | 13 | 2205 | |||
XM_008761087.1 | XP_008759309.1 | 12 | 1347 | |||
XM_008761088.1 | XP_008759310.1 | 12 | 1279 | |||
GenBank mRNA | AY509976.1 | - | 16 | 1724 |
