siRNA Details
| Gene Symbol: Rxfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201417.1 | NP_958820.1 | 17, 18 | 1969 | ||
| XM_006232508.2 | XP_006232570.1 | 17, 18 | 2106 | |||
| XM_006232510.2 | XP_006232572.1 | 14, 15 | 1660 | |||
| XM_008761085.1 | XP_008759307.1 | 13, 14 | 3410 | |||
| XM_008761086.1 | XP_008759308.1 | 14, 15 | 2450 | |||
| XM_008761087.1 | XP_008759309.1 | 13, 14 | 1592 | |||
| XM_008761088.1 | XP_008759310.1 | 13, 14 | 1524 | |||
| GenBank mRNA | AY509976.1 | - | 17, 18 | 1969 | ||

Rxfp1
Genomic Map