siRNA Details
| Gene Symbol: Mnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232551.2 | XP_006232613.1 | 6 | 578 | ||
| XM_008761117.1 | XP_008759339.1 | 6 | 580 | |||
| XM_215595.7 | XP_215595.5 | 5 | 658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mnd1| Gene Symbol: Mnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232551.2 | XP_006232613.1 | 6 | 578 | ||
| XM_008761117.1 | XP_008759339.1 | 6 | 580 | |||
| XM_215595.7 | XP_215595.5 | 5 | 658 | |||