siRNA Details
Gene Symbol: LOC102550932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232552.2 | XP_006232614.1 | 1 | 2868 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102550932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232552.2 | XP_006232614.1 | 1 | 2868 |