siRNA Details
| Gene Symbol: Sh3d19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006232783.2 | XP_006232845.1 | 7 | 1321 | ||
| XM_006232784.2 | XP_006232846.1 | 8 | 1666 | |||
| XM_006232785.2 | XP_006232847.1 | 7 | 1323 | |||
| XM_215597.8 | XP_215597.6 | 7 | 1319 | |||

Sh3d19
Genomic Map