siRNA Details
| Gene Symbol: Padi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017230.2 | NP_058926.1 | 7 | 711 | ||
| GenBank mRNA | D88034.1 | - | 7 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Padi3| Gene Symbol: Padi3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017230.2 | NP_058926.1 | 7 | 711 | ||
| GenBank mRNA | D88034.1 | - | 7 | 712 | ||