siRNA Details
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024979.1 | NP_001020150.1 | 3 | 248 | ||
XM_008761129.1 | XP_008759351.1 | 4 | 779 | |||
XM_008761130.1 | XP_008759352.1 | 4 | 829 | |||
XM_008761131.1 | XP_008759353.1 | 3 | 765 | |||
XM_008761132.1 | XP_008759354.1 | 4 | 943 | |||
GenBank mRNA | BC093381.1 | - | 3 | 248 | 7388663 |
