siRNA Details
| Gene Symbol: Cct3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199091.2 | NP_954522.1 | 11 | 1059 | ||
| GenBank mRNA | BC063178.1 | - | 11 | 1063 | 6890550 | |
| FQ213900.1 | - | 4 | 301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct3| Gene Symbol: Cct3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199091.2 | NP_954522.1 | 11 | 1059 | ||
| GenBank mRNA | BC063178.1 | - | 11 | 1063 | 6890550 | |
| FQ213900.1 | - | 4 | 301 | |||