siRNA Details
Gene Symbol: Cct3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199091.2 | NP_954522.1 | 7 | 588 | ||
GenBank mRNA | BC063178.1 | - | 7 | 592 | 6890550 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cct3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199091.2 | NP_954522.1 | 7 | 588 | ||
GenBank mRNA | BC063178.1 | - | 7 | 592 | 6890550 |