siRNA Details
| Gene Symbol: Ssr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106442.1 | NP_001099912.1 | 2 | 161 | ||
| GenBank mRNA | BC168754.1 | - | 2 | 133 | 6886830 | |
| FQ213377.1 | - | 2 | 125 | |||
| FQ221799.1 | - | 2 | 99 | |||
| FQ229540.1 | - | 2 | 133 | |||
| FQ232929.1 | - | 2 | 117 | |||
| FQ235190.1 | - | 2 | 117 | |||

Ssr2
Genomic Map