siRNA Details
| Gene Symbol: Dap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011950.2 | NP_001011950.2 | 11 | 1218 | ||
| GenBank mRNA | BC085878.1 | - | 11 | 1218 | 7191747 | |
| FQ230674.1 | - | 11 | 1007 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dap3| Gene Symbol: Dap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011950.2 | NP_001011950.2 | 11 | 1218 | ||
| GenBank mRNA | BC085878.1 | - | 11 | 1218 | 7191747 | |
| FQ230674.1 | - | 11 | 1007 | |||