siRNA Details
| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106448.2 | NP_001099918.1 | 7 | 940 | ||
| GenBank mRNA | BC166534.1 | - | 7 | 940 | 7190922 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2q1| Gene Symbol: Ube2q1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106448.2 | NP_001099918.1 | 7 | 940 | ||
| GenBank mRNA | BC166534.1 | - | 7 | 940 | 7190922 | |