siRNA Details
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017231.1 | NP_058927.1 | 3, 4 | 402 | ||
GenBank mRNA | BC070945.1 | - | 3, 4 | 430 | 7103390 | |
M25758.1 | - | 3, 4 | 402 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017231.1 | NP_058927.1 | 3, 4 | 402 | ||
GenBank mRNA | BC070945.1 | - | 3, 4 | 430 | 7103390 | |
M25758.1 | - | 3, 4 | 402 |