siRNA Details
| Gene Symbol: Man1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106452.1 | NP_001099922.1 | 7 | 1420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man1a2| Gene Symbol: Man1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106452.1 | NP_001099922.1 | 7 | 1420 | ||