siRNA Details
Gene Symbol: Hpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017233.1 | NP_058929.1 | 10 | 733 | ||
GenBank mRNA | AF082834.1 | - | 10 | 733 | ||
BC081819.1 | - | 10 | 731 | 7107086 | ||
FQ210905.1 | - | 10 | 741 | |||
FQ218378.1 | - | 10 | 742 | |||
FQ218745.1 | - | 10 | 740 | |||
FQ219696.1 | - | 10 | 741 | |||
M18405.1 | - | 9 | 689 |
