siRNA Details
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013913.1 | NP_001013935.1 | 5 | 688 | ||
XM_008761384.1 | XP_008759606.1 | 5 | 859 | |||
XM_008761385.1 | XP_008759607.1 | 5 | 862 | |||
XM_008761386.1 | XP_008759608.1 | 5 | 857 | |||
XM_008761387.1 | XP_008759609.1 | 5 | 857 | |||
XM_008761388.1 | XP_008759610.1 | 5 | 863 | |||
XM_008761389.1 | XP_008759611.1 | 5 | 851 | |||
XM_008761390.1 | XP_008759612.1 | 5 | 852 | |||
GenBank mRNA | BC085920.1 | - | 5 | 688 | 7108742 |
