siRNA Details
| Gene Symbol: LOC100912384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773861.1 | XP_008772083.1 | 8 | 955 | ||
| Gene Symbol: Eps8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106463.1 | NP_001099933.1 | 8 | 801 | ||

LOC100912384
Genomic Map