siRNA Details
| Gene Symbol: Vav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191714.1 | NP_001178643.1 | 6 | 631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vav3| Gene Symbol: Vav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191714.1 | NP_001178643.1 | 6 | 631 | ||