siRNA Details
| Gene Symbol: Ints12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007640.1 | NP_001007641.1 | 6 | 1013 | ||
| XM_006233320.2 | XP_006233382.1 | 7 | 2143 | |||
| XM_006233321.2 | XP_006233383.1 | 7 | 1129 | |||
| GenBank mRNA | BC079404.1 | - | 6 | 1013 | 7112817 | |
| FQ212179.1 | - | 7 | 1104 | |||

Ints12
Genomic Map