siRNA Details
| Gene Symbol: Tspyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013033.1 | NP_001013051.1 | 1 | 755 | ||
| GenBank mRNA | BC081955.1 | - | 1 | 755 | 7123128 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspyl1| Gene Symbol: Tspyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013033.1 | NP_001013051.1 | 1 | 755 | ||
| GenBank mRNA | BC081955.1 | - | 1 | 755 | 7123128 | |