siRNA Details
| Gene Symbol: Bdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106473.1 | NP_001099943.1 | 5 | 493 | ||
| XM_006233322.2 | XP_006233384.1 | 5 | 402 | |||
| GenBank mRNA | BC095848.1 | - | 4 | 887 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bdh2| Gene Symbol: Bdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106473.1 | NP_001099943.1 | 5 | 493 | ||
| XM_006233322.2 | XP_006233384.1 | 5 | 402 | |||
| GenBank mRNA | BC095848.1 | - | 4 | 887 | ||