siRNA Details
| Gene Symbol: Sec24b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106474.1 | NP_001099944.1 | 5 | 1323 | ||
| XM_006233275.2 | XP_006233337.1 | 5 | 1399 | |||
| XM_006233276.2 | XP_006233338.1 | 4 | 1293 | |||
| XM_006233277.2 | XP_006233339.1 | 3 | 1119 | |||
| XM_008761458.1 | XP_008759680.1 | 5 | 1399 | |||

Sec24b
Genomic Map