siRNA Details
| Gene Symbol: Sec24b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106474.1 | NP_001099944.1 | 3, 4 | 1177 | ||
| XM_006233275.2 | XP_006233337.1 | 3, 4 | 1253 | |||
| XM_008761458.1 | XP_008759680.1 | 3, 4 | 1253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec24b| Gene Symbol: Sec24b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106474.1 | NP_001099944.1 | 3, 4 | 1177 | ||
| XM_006233275.2 | XP_006233337.1 | 3, 4 | 1253 | |||
| XM_008761458.1 | XP_008759680.1 | 3, 4 | 1253 | |||