siRNA Details
| Gene Symbol: Adh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106475.1 | NP_001099945.1 | 3 | 399 | ||
| XM_006233371.2 | XP_006233433.1 | 4 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adh6a| Gene Symbol: Adh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106475.1 | NP_001099945.1 | 3 | 399 | ||
| XM_006233371.2 | XP_006233433.1 | 4 | 481 | |||