siRNA Details
| Gene Symbol: Tnni3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181769.1 | NP_861434.1 | 11 | 1114 | ||
| XM_008761563.1 | XP_008759785.1 | 11 | 1278 | |||
| GenBank mRNA | AY303692.1 | - | 11 | 1114 | ||
| BC098648.1 | - | 11 | 1119 | |||

Tnni3k
Genomic Map