siRNA Details
Gene Symbol: Tnni3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181769.1 | NP_861434.1 | 19 | 1896 | ||
GenBank mRNA | AY303692.1 | - | 19 | 1896 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnni3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181769.1 | NP_861434.1 | 19 | 1896 | ||
GenBank mRNA | AY303692.1 | - | 19 | 1896 |