siRNA Details
| Gene Symbol: Kcnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033376.1 | NP_203694.1 | 2 | 463 | ||
| GenBank mRNA | AB048823.1 | - | 3 | 316 | ||
| AB048824.1 | - | 3 | 305 | |||
| AF031384.1 | - | 2 | 463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk3| Gene Symbol: Kcnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033376.1 | NP_203694.1 | 2 | 463 | ||
| GenBank mRNA | AB048823.1 | - | 3 | 316 | ||
| AB048824.1 | - | 3 | 305 | |||
| AF031384.1 | - | 2 | 463 | |||