siRNA Details
| Gene Symbol: Grik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111114.1 | NP_001104584.1 | 3 | 673 | ||
| NM_001111117.1 | NP_001104587.1 | 3 | 673 | |||
| NM_017241.2 | NP_058937.1 | 3 | 673 | |||
| XM_006248053.2 | XP_006248115.1 | 3 | 1057 | |||
| XM_006248054.2 | XP_006248116.1 | 3 | 1057 | |||
| GenBank mRNA | M83560.1 | - | 3 | 675 | ||
| M83561.1 | - | 3 | 675 | |||
| Z11712.1 | - | 3 | 519 | |||
| Z11713.1 | - | 3 | 519 | |||
| Z11714.1 | - | 3 | 519 | |||

Grik1
Genomic Map