siRNA Details
| Gene Symbol: Ermn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008311.3 | NP_001008312.1 | 1 | 290 | ||
| GenBank mRNA | BC086596.1 | - | 1 | 290 | 7311836 | |
| DQ119821.1 | - | 218 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ermn| Gene Symbol: Ermn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008311.3 | NP_001008312.1 | 1 | 290 | ||
| GenBank mRNA | BC086596.1 | - | 1 | 290 | 7311836 | |
| DQ119821.1 | - | 218 | ||||