siRNA Details
| Gene Symbol: Eef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017245.2 | NP_058941.1 | 9 | 1997 | ||
| GenBank mRNA | AF000576.1 | - | 1 | 289 | ||
| BC066661.1 | - | 9 | 1997 | 5623322 | ||
| FQ215372.1 | - | 1 | 241 | |||
| FQ220112.1 | - | 1 | 233 | |||
| FQ229435.1 | - | 1 | 51 | |||
| K03502.1 | - | 2 | 391 | |||
| Y07504.1 | - | 9 | 1983 | |||

Eef2
Genomic Map