siRNA Details
Gene Symbol: Nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004229.1 | NP_001004229.1 | 2 | 204 | ||
XM_006234430.2 | XP_006234492.1 | 2 | 211 | |||
GenBank mRNA | BC078914.1 | - | 2 | 204 | 7126643 |
