siRNA Details
| Gene Symbol: Nup35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004229.1 | NP_001004229.1 | 5 | 540 | ||
| XM_006234430.2 | XP_006234492.1 | 5 | 547 | |||
| GenBank mRNA | BC078914.1 | - | 5 | 540 | 7126643 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup35| Gene Symbol: Nup35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004229.1 | NP_001004229.1 | 5 | 540 | ||
| XM_006234430.2 | XP_006234492.1 | 5 | 547 | |||
| GenBank mRNA | BC078914.1 | - | 5 | 540 | 7126643 | |