siRNA Details
| Gene Symbol: Olr477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000304.1 | NP_001000304.1 | 1 | 69 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr477| Gene Symbol: Olr477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000304.1 | NP_001000304.1 | 1 | 69 | ||