siRNA Details
Gene Symbol: Esyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017249.2 | NP_058945.2 | 6 | 964 | ||
GenBank mRNA | AF099138.1 | - | 6 | 909 | ||
BC101857.1 | - | 6 | 964 | 7937926 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Esyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017249.2 | NP_058945.2 | 6 | 964 | ||
GenBank mRNA | AF099138.1 | - | 6 | 909 | ||
BC101857.1 | - | 6 | 964 | 7937926 |