siRNA Details
| Gene Symbol: Mtch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106488.2 | NP_001099958.2 | 6 | 468 | ||
| GenBank mRNA | BC158687.1 | - | 6 | 419 | 8375010 | |
| DY311538.1 | - | 419 | ||||
| EV779017.1 | - | 489 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtch2| Gene Symbol: Mtch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106488.2 | NP_001099958.2 | 6 | 468 | ||
| GenBank mRNA | BC158687.1 | - | 6 | 419 | 8375010 | |
| DY311538.1 | - | 419 | ||||
| EV779017.1 | - | 489 | ||||