siRNA Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039196.1 | NP_001034285.1 | 2 | 311 | ||
| XM_006234501.2 | XP_006234563.1 | 2 | 517 | |||
| XM_006234502.2 | XP_006234564.1 | 2 | 521 | |||
| GenBank mRNA | BC112321.1 | - | 2 | 311 | 7933412 | |

Slc39a13
Genomic Map