siRNA Details
Gene Symbol: Apip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106492.1 | NP_001099962.1 | 6 | 675 | ||
XM_006234638.2 | XP_006234700.1 | 6 | 724 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Apip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106492.1 | NP_001099962.1 | 6 | 675 | ||
XM_006234638.2 | XP_006234700.1 | 6 | 724 |