siRNA Details
| Gene Symbol: Ifrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019242.1 | NP_062115.1 | 2 | 249 | ||
| XM_008764643.1 | XP_008762865.1 | 1 | 868 | |||
| GenBank mRNA | BC083560.1 | - | 2 | 315 | 7193329 | |
| J04511.1 | - | 2 | 249 | |||

Ifrd1
Genomic Map