siRNA Details
| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005871.1 | NP_001005871.1 | 7, 8 | 1267 | ||
| XM_008769446.1 | XP_008767668.1 | 8, 9 | 1650 | |||
| XM_008769447.1 | XP_008767669.1 | 8, 9 | 2287 | |||
| XM_008769448.1 | XP_008767670.1 | 7, 8 | 1614 | |||
| XM_008769449.1 | XP_008767671.1 | 8, 9 | 1651 | |||
| XM_008769450.1 | XP_008767672.1 | 7, 8 | 1615 | |||
| GenBank mRNA | U15408.1 | - | 7, 8 | 1267 | ||
| X76452.1 | - | 3, 4 | 306 | |||

Atp2b4
Genomic Map