siRNA Details
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106494.1 | NP_001099964.1 | 10 | 1383 | ||
XM_006234702.2 | XP_006234764.1 | 10 | 1405 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106494.1 | NP_001099964.1 | 10 | 1383 | ||
XM_006234702.2 | XP_006234764.1 | 10 | 1405 |