siRNA Details
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106497.1 | NP_001099967.1 | 4 | 301 | ||
GenBank mRNA | BC166931.1 | - | 4 | 257 | 6921277 | |
FQ210704.1 | - | 3 | 201 | |||
FQ210780.1 | - | 3 | 148 |
