siRNA Details
| Gene Symbol: Usp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106502.1 | NP_001099972.1 | 13 | 2181 | ||
| XM_006234916.2 | XP_006234978.1 | 13 | 2254 | |||
| XM_008762155.1 | XP_008760377.1 | 13 | 2254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp8| Gene Symbol: Usp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106502.1 | NP_001099972.1 | 13 | 2181 | ||
| XM_006234916.2 | XP_006234978.1 | 13 | 2254 | |||
| XM_008762155.1 | XP_008760377.1 | 13 | 2254 | |||