siRNA Details
| Gene Symbol: Tmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106510.1 | NP_001099980.1 | 14 | 1975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmc2| Gene Symbol: Tmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106510.1 | NP_001099980.1 | 14 | 1975 | ||