siRNA Details
| Gene Symbol: Vps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005541.1 | NP_001005541.1 | 17 | 1669 | ||
| GenBank mRNA | BC081963.1 | - | 17 | 1669 | 7124985 | |
| FQ212033.1 | - | 14 | 1975 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps16| Gene Symbol: Vps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005541.1 | NP_001005541.1 | 17 | 1669 | ||
| GenBank mRNA | BC081963.1 | - | 17 | 1669 | 7124985 | |
| FQ212033.1 | - | 14 | 1975 | |||